Learn More
Aat Bioquest Nuclear Green LCS1 5 mM DMSO Solution (0.5 ml)
SDP
Catalog No. 502249735
Encompass_Preferred
Our Nuclear Green LCS1 is a fluorogenic DNA-selective and cell-permeant dye for analyzing DNA content in living cells. This catalog is for 0.5 ml.
This item is not returnable. View return policy
Catalog No. 50-224-9735 Supplier Aat Bioquest Supplier No. 17540
Our Nuclear Green LCS1 is a fluorogenic DNA-selective and cell-permeant dye for analyzing DNA content in living cells. This catalog is for 0.5 ml.