Learn More
Enzo Life Sciences NUCLEAR-ID Green chromatin condensation detection kit (1Kit)
SDP
Catalog No. 502012593
Encompass_Preferred
The NUCLEAR-ID Green Chromatin Condensation Detection Kit detects nuclear condensation using a DNA intercalating dye that brightly stains the condensed chromatin of apoptotic cells, but only dimly stains the uncondensed chromatin of healthy living cells. Mechanism of Action The cell-permeable dye used in this application is an aromatic, planar cationic structure that inserts between stacked base pairs on the DNA duplex, providing an environmentally-dependent fluorescence enhancement of the dye molecules and large increases in fluorescence signal relative to the free dye in solution. Since the signal enhancement provides a proportional response, direct quantitation of DNA is possible. Further signal increase is observed upon DNA condensation during apoptosis.Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2593 Supplier Enzo Life Sciences Supplier No. ENZ51021K200
The NUCLEAR-ID Green Chromatin Condensation Detection Kit detects nuclear condensation using a DNA intercalating dye that brightly stains the condensed chromatin of apoptotic cells, but only dimly stains the uncondensed chromatin of healthy living cells. Mechanism of Action The cell-permeable dye used in this application is an aromatic, planar cationic structure that inserts between stacked base pairs on the DNA duplex, providing an environmentally-dependent fluorescence enhancement of the dye molecules and large increases in fluorescence signal relative to the free dye in solution. Since the signal enhancement provides a proportional response, direct quantitation of DNA is possible. Further signal increase is observed upon DNA condensation during apoptosis.Applications: Flow Cytometry, Fluorescence microscopy, Fluorescent detection .