Learn More
Enzo Life Sciences NUCLEAR-ID Red/Green cell viability reagent, (100µl)
SDP
Catalog No. NC1292237
Encompass_Preferred
Distinguishes between live vs dead cells in a single reagent. The NUCLEAR-ID Red/Green cell viability reagent (Prod. No. ENZ-53006) is a mixture of a red fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers. Purity: ≥93% (HPLC). Long Term Storage: -80°C
Catalog No. NC1292237 Supplier Enzo Life Sciences Supplier No. ENZ53006C100
Distinguishes between live vs dead cells in a single reagent. The NUCLEAR-ID Red/Green cell viability reagent (Prod. No. ENZ-53006) is a mixture of a red fluorescent cell-permeable nucleic acid dye and a green fluorescent cell-impermeable nucleic acid dye that is suited for staining dead nuclei. The staining pattern arising from the simultaneous combination of these two dyes permits determination of live and dead cell populations by fluorescence/confocal microscopy. The reagent, supplied as a 1000x solution, is sufficient for 1000 microscopy assays. The single-tube format makes this cell viability reagent easy to use. It leaves the cytoplasm unstained, potentially allowing visualization of other cell markers. Purity: ≥93% (HPLC). Long Term Storage: -80°C