Learn More
Cusabio Technology LLC Human Nuclear matrix protein 22,NMP-22 ELISA Kit
Catalog No. NC2952883
Encompass_Preferred
This Human NMP-22 ELISA Kit was designed for the quantitative measurement of Human NMP-22 protein in serum, urine. It is a Competitive ELISA kit, its detection range is 0.156 ng/mL-40 ng/mL and the sensitivity is 0.156 ng/mL. One kit contains 96 wells.
Catalog No. NC2952883 Supplier Cusabio Technology LLC Supplier No. CSBE09037H
This Human NMP-22 ELISA Kit was designed for the quantitative measurement of Human NMP-22 protein in serum, urine. It is a Competitive ELISA kit, its detection range is 0.156 ng/mL-40 ng/mL and the sensitivity is 0.156 ng/mL. One kit contains 96 wells.