Learn More
Aat Bioquest Nuclear Orange LCS1 5 mM DMSO Solution (0.5 ml)
SDP
Catalog No. 502249736
Encompass_Preferred
Our Nuclear Orange LCS1 is a fluorogenic DNA-selective and cell-permeant dye for analyzing DNA content in living cells. This catalog is for 0.5 ml.
This item is not returnable. View return policy
Catalog No. 50-224-9736 Supplier Aat Bioquest Supplier No. 17541
Our Nuclear Orange LCS1 is a fluorogenic DNA-selective and cell-permeant dye for analyzing DNA content in living cells. This catalog is for 0.5 ml.