Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Nuclear Receptor Buffer F

Catalog No. 502576139
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-257-6139 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-257-6139 Supplier Invitrogen™ Supplier No. PV4547
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Nuclear Receptor Buffer F

A specialized nuclear receptor buffer that can be used with Invitrogen's collection of purified nuclear receptor proteins and assays.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nuclear Receptor Buffer
Physical Form Liquid
Quantity 25 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.