Learn More
Ethos Biosciences Classical Harris Hematoxylin, 5gal, Ethos
Catalog No. 5000574110
Encompass
Classical Acidified Harris Hematoxylin Formulation is a nuclear stain designed for routine histology and cytology applications. This traditional formulation incorporates controlled acidification to support crisp nuclear detail and predictable differentiation in hematoxylin and eosin (H&E) procedures. It provides strong, well-defined blue to purple nuclear staining when followed by appropriate bluing steps. Compatible with manual and automated staining systems, the formulation delivers consistent performance and reproducible results across paraffin and frozen tissue sections. Suitable for clinical, research, and educational laboratories requiring reliable routine nuclear staining.
Catalog No. 50-005-74110 Supplier Ethos Biosciences Supplier No. 70025G
Classical Acidified Harris Hematoxylin Formulation is a nuclear stain designed for routine histology and cytology applications. This traditional formulation incorporates controlled acidification to support crisp nuclear detail and predictable differentiation in hematoxylin and eosin (H&E) procedures. It provides strong, well-defined blue to purple nuclear staining when followed by appropriate bluing steps. Compatible with manual and automated staining systems, the formulation delivers consistent performance and reproducible results across paraffin and frozen tissue sections. Suitable for clinical, research, and educational laboratories requiring reliable routine nuclear staining.