Learn More
Ethos Biosciences Modified Harris Hematoxylin, 1gal, Ethos
Catalog No. 5000574260
Encompass_Preferred
Harris Hematoxylin is a nuclear stain designed for routine histology and cytology applications. This accelerated formulation provides crisp, well-defined nuclear detail with reduced staining time, supporting efficient laboratory workflows. Suitable for use in H&E staining protocols, it delivers strong nuclear coloration and consistent performance across paraffin and frozen tissue sections. Compatible with standard differentiation and bluing steps, the stain supports reliable, reproducible results in clinical, research, and educational laboratories. Intended for professional laboratory use and integration into manual or automated staining systems.
Catalog No. 50-005-74260 Supplier Ethos Biosciences Supplier No. 7000G
Harris Hematoxylin is a nuclear stain designed for routine histology and cytology applications. This accelerated formulation provides crisp, well-defined nuclear detail with reduced staining time, supporting efficient laboratory workflows. Suitable for use in H&E staining protocols, it delivers strong nuclear coloration and consistent performance across paraffin and frozen tissue sections. Compatible with standard differentiation and bluing steps, the stain supports reliable, reproducible results in clinical, research, and educational laboratories. Intended for professional laboratory use and integration into manual or automated staining systems.