Learn More
Abcam Bisbenzimide H 33342 (Hoechst 33342), adenine-thymine selective DNA fluorescent stain, 100MG
Catalog No. NC1962822
Encompass_Preferred
MW 561.9 Da, Purity >98%. Cell-permeable, adenine-thymine selective DNA fluorescent stain. DNA minor groove-directed agent. Labels DNA, chromosomes and nuclei. Stimulates topoisomerase I-mediated DNA cleavage. Bisbenzimide H 33258 (ab145596) analog.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1962822 Supplier Abcam Supplier No. AB145597100MG
MW 561.9 Da, Purity >98%. Cell-permeable, adenine-thymine selective DNA fluorescent stain. DNA minor groove-directed agent. Labels DNA, chromosomes and nuclei. Stimulates topoisomerase I-mediated DNA cleavage. Bisbenzimide H 33258 (ab145596) analog.
The product is subject to the following: Abcam Restricted Use Statement