Learn More
Aat Bioquest Nuclear Violet DCS1 5 mM DMSO Solution (0.5 ml)
SDP
Catalog No. 502249740
Encompass_Preferred
Our Nuclear Violet DCS1 is a fluorogenic DNA-selective and live cell-impermeant dye for analyzing DNA content in dead or fixed cells. This catalog is for 0.5 ml.
This item is not returnable. View return policy
Catalog No. 50-224-9740 Supplier Aat Bioquest Supplier No. 17549
Our Nuclear Violet DCS1 is a fluorogenic DNA-selective and live cell-impermeant dye for analyzing DNA content in dead or fixed cells. This catalog is for 0.5 ml.