Learn More
Cayman Chemical Nuclear Yelow 25mg
SDP
Supplier:  Cayman Chemical 2517825 MG
Encompass_Preferred
A fluorescent DNA probe for live and fixed cells; fluoresces upon binding to double-stranded DNA in live or fixed cells; has commonly been used in combination with retrograde tracers for two-color neuronal mapping; ex/em = 355/495 nm
Catalog No. 50-293-4663