Learn More
Cayman Chemical Nuclear Yelow 5mg
SDP
Catalog No. 502934765
Encompass_Preferred
A fluorescent DNA probe for live and fixed cells; fluoresces upon binding to double-stranded DNA in live or fixed cells; has commonly been used in combination with retrograde tracers for two-color neuronal mapping; ex/em = 355/495 nm
Catalog No. 50-293-4765 Supplier Cayman Chemical Supplier No. 251785 MG
A fluorescent DNA probe for live and fixed cells; fluoresces upon binding to double-stranded DNA in live or fixed cells; has commonly been used in combination with retrograde tracers for two-color neuronal mapping; ex/em = 355/495 nm