Learn More
Sino Biological Nuclease Activity QC System 96 Tests
Catalog No. 5000696261
Encompass_Preferred
The Nuclease Activity QC System offers a stable, direct, and user-friendly solution for nuclease qualification, utilizing fluorescent hairpin DNA substrates.
Catalog No. 50-006-96261 Supplier Sino Biological Supplier No. NE00196
Add to Cart
Add to Cart
The Nuclease Activity QC System offers a stable, direct, and user-friendly solution for nuclease qualification, utilizing fluorescent hairpin DNA substrates.