Learn More
New England Biolabs, Inc. Nuclease-Free Water - 500 ml
SDP
Catalog No. 5000460442
Encompass
Nuclease-free Water is ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in the production of this water, so as to avoid inhibition in enzymatic reactions.
  • Ideal for the preparation of reagents and for use in enzymatic reactions
  • No toxic agents, such as DEPC, are used in production, so as to avoid inhibition in enzymatic rxns
  • Available in 25 ml and 100 ml sizes
Catalog No. 50-004-60442 Supplier New England Biolabs, Inc. Supplier No. B1500X
Nuclease-free Water is ideal for the preparation of reagents and for use in enzymatic reactions. No toxic agents, such as DEPC, are used in the production of this water, so as to avoid inhibition in enzymatic reactions.
  • Ideal for the preparation of reagents and for use in enzymatic reactions
  • No toxic agents, such as DEPC, are used in production, so as to avoid inhibition in enzymatic rxns
  • Available in 25 ml and 100 ml sizes