Learn More
STEMCELL Technologies Nuclease-Free Water, Size: 20 mL
SDP
Catalog No. 501976787
Encompass
Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.
Catalog No. 50-197-6787 Supplier STEMCELL Technologies Supplier No. 79002.1
Nuclease-Free Water is used in various molecular biology applications requiring nuclease-free conditions, such as in processing DNA or RNA (i.e. PCR, cDNA synthesis, or qPCR). This product is sterile-filtered (0.2 µm), free of RNase and DNase activity, endotoxin-free, and not DEPC-treated.