Learn More
Worthington Biochemical Corporation Miccrococal Nuclease, 45ku
SDP
Catalog No. NC9391488
Encompass
Nuclease, Micrococcal (S7); Micrococcal nuclease (MN) catalyzes cleavage of both DNA and RNA to yield 3'-nucleotides; MN is the extracellular nuclease of Staphylococcus aureus; M.W. 16807; Optimum 9.2 pH; Stored: 2 deg. to 8 deg.C
Catalog No. NC9391488 Supplier Worthington Biochemical Corporation Supplier No. LS004798
Nuclease, Micrococcal (S7); Micrococcal nuclease (MN) catalyzes cleavage of both DNA and RNA to yield 3'-nucleotides; MN is the extracellular nuclease of Staphylococcus aureus; M.W. 16807; Optimum 9.2 pH; Stored: 2 deg. to 8 deg.C