Learn More
Sigma Aldrich Fine Chemicals Biosciences Nuclease P1 from Penicillium citrinum, 54576-84-0, MFCD00131010, 1VL
Catalog No. NC1553657
Encompass
Lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase), Synonym: Endonuclease P1. Nuclease P1 from Penicillium citrinum has been used in a study to assess crystal structures using ammonium sulphate or polyethylene glycol 4000 as a precipitating agent. It has also been used in a study to investigate a method for the direct sequence analysis 20-25 nucleotides from the terinini of 5′ or 3′ end group labeled RNA. Nuclease P1 is used to improve the sensitivity of a 32P-labeling method for the detection of DNA adducts.
Catalog No. NC1553657 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N86301VL
Lyophilized powder, ≥200 units/mg protein (E1%/280, 3′-5′-Phosphodiesterase), Synonym: Endonuclease P1. Nuclease P1 from Penicillium citrinum has been used in a study to assess crystal structures using ammonium sulphate or polyethylene glycol 4000 as a precipitating agent. It has also been used in a study to investigate a method for the direct sequence analysis 20-25 nucleotides from the terinini of 5′ or 3′ end group labeled RNA. Nuclease P1 is used to improve the sensitivity of a 32P-labeling method for the detection of DNA adducts.