Learn More
New England Biolabs, Inc. Nuclease P1 – 10000 units
SDP
Catalog No. 501959360
Encompass
Nuclease P1 (from P. citrinum) is a zinc-dependent single-strand specific nuclease which hydrolyzes 3->5 phosphodiester bonds in RNA and ssDNA with no base specificity. Nuclease P1 also exhibits 3-phosphomonoesterase activity. Although a single-strand specific nuclease (ssDNA and RNA-specific), Nuclease P1 does display some activity toward dsDNA in Nuclease P1 Reaction Buffer. If preferentially degrading single-stranded nucleic acids (ssDNA or RNA) in the presence of double stranded DNA (dsDNA), we recommend using Nuclease P1 in 1X NEBuffer r1.1 to limit activity on dsDNA while maintaining single-strand nuclease activity.
Catalog No. 50-195-9360 Supplier New England Biolabs, Inc. Supplier No. M0660S
Nuclease P1 (from P. citrinum) is a zinc-dependent single-strand specific nuclease which hydrolyzes 3->5 phosphodiester bonds in RNA and ssDNA with no base specificity. Nuclease P1 also exhibits 3-phosphomonoesterase activity. Although a single-strand specific nuclease (ssDNA and RNA-specific), Nuclease P1 does display some activity toward dsDNA in Nuclease P1 Reaction Buffer. If preferentially degrading single-stranded nucleic acids (ssDNA or RNA) in the presence of double stranded DNA (dsDNA), we recommend using Nuclease P1 in 1X NEBuffer r1.1 to limit activity on dsDNA while maintaining single-strand nuclease activity.