Learn More
Medchemexpress LLC Nuclease P1 | 54576-84-0 | ≥200 U/mg protein | 250 U
SDP
Catalog No. 5000397209
Encompass_Preferred
Nuclease P1 is a single-stranded specific endonuclease that hydrolyzes nucleic acids into 5'-mononucleotides and cleaves the single-stranded region of a double-stranded nucleic acid. It is obtained by fermentation of *Penicillium citrinum* through extraction, ultrafiltration concentration, drying, and purification processes. It is widely used in the pharmaceutical and food industries.
  • Hydrolyzes nucleic acids into 5'-mononucleotides
  • Cleaves the single-stranded region of a double-stranded nucleic acid
  • Used in the pharmaceutical and food industries
Catalog No. 50-003-97209 Supplier Medchemexpress LLC Supplier No. 250UHYP2963
Nuclease P1 is a single-stranded specific endonuclease that hydrolyzes nucleic acids into 5'-mononucleotides and cleaves the single-stranded region of a double-stranded nucleic acid. It is obtained by fermentation of *Penicillium citrinum* through extraction, ultrafiltration concentration, drying, and purification processes. It is widely used in the pharmaceutical and food industries.
  • Hydrolyzes nucleic acids into 5'-mononucleotides
  • Cleaves the single-stranded region of a double-stranded nucleic acid
  • Used in the pharmaceutical and food industries