Learn More
Medchemexpress LLC Nuclease S1, Aspergillus oryzae | 37288-25-8 | 10 KU
SDP
Catalog No. 5000397348
Encompass_Preferred
Nuclease S1, Aspergillus oryzae is a specific endonuclease that degrades single-stranded DNA (ssDNA) and RNA. It can also remove protruding single-strand ends from double-stranded DNA. This product is for research use only.
  • Specific endonuclease that degrades single-stranded DNA (ssDNA) and RNA
  • Removes protruding single-strand ends from double-stranded DNA
  • Sourced from Aspergillus oryzae
  • Enzyme activity: ≥100000 U/mL
  • Formulated in 30mM sodium acetate, pH 4.6, 50mM NaCl, 1mM ZnCl2, 50% glycerol
Catalog No. 50-003-97348 Supplier Medchemexpress LLC Supplier No. HYP277310KU
Nuclease S1, Aspergillus oryzae is a specific endonuclease that degrades single-stranded DNA (ssDNA) and RNA. It can also remove protruding single-strand ends from double-stranded DNA. This product is for research use only.
  • Specific endonuclease that degrades single-stranded DNA (ssDNA) and RNA
  • Removes protruding single-strand ends from double-stranded DNA
  • Sourced from Aspergillus oryzae
  • Enzyme activity: ≥100000 U/mL
  • Formulated in 30mM sodium acetate, pH 4.6, 50mM NaCl, 1mM ZnCl2, 50% glycerol