Learn More
Sigma Aldrich Fine Chemicals Biosciences Nuclease S1 from Aspergill
Catalog No. 502901956
Encompass
The Nuclease S1 enzyme from Aspergillus oryzae has the ability to degrade single-stranded oligonucleotides composed of either deoxynucleotides or ribonucleotides.
Catalog No. 50-290-1956 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. N566150KU
The Nuclease S1 enzyme from Aspergillus oryzae has the ability to degrade single-stranded oligonucleotides composed of either deoxynucleotides or ribonucleotides.