Learn More
Medchemexpress LLC Nuclease, Serratia marcescens | 9025-65-4 | 95% | 25 KU
SDP
Catalog No. 5000342810
Encompass_Preferred
Serratia marcescens nuclease (EC 3.1.30.2) is a nonspecific nuclease with broad utility due to its potent digestive activity toward both DNA and RNA. It is an endonuclease that can completely digest all forms of DNA and RNA (including single-stranded, double-stranded, linear and circular) into 5'-monophosphate oligonucleotides of 3-5 bases in length. It has no protein activity and is highly specific. It is expressed in a yeast system and is suitable for the removal of nucleic acids in various proteins and biological products. The reaction conditions are mild, the reaction time is short, and the nucleic acid removal efficiency is high.
  • No protein activity
  • Highly specific
  • Expressed in a yeast system
  • Suitable for the removal of nucleic acids in various proteins and biological products
  • Mild reaction conditions
  • Short reaction time
  • High nucleic acid removal efficiency
Catalog No. 50-003-42810 Supplier Medchemexpress LLC Supplier No. 25KUHY131160
Serratia marcescens nuclease (EC 3.1.30.2) is a nonspecific nuclease with broad utility due to its potent digestive activity toward both DNA and RNA. It is an endonuclease that can completely digest all forms of DNA and RNA (including single-stranded, double-stranded, linear and circular) into 5'-monophosphate oligonucleotides of 3-5 bases in length. It has no protein activity and is highly specific. It is expressed in a yeast system and is suitable for the removal of nucleic acids in various proteins and biological products. The reaction conditions are mild, the reaction time is short, and the nucleic acid removal efficiency is high.
  • No protein activity
  • Highly specific
  • Expressed in a yeast system
  • Suitable for the removal of nucleic acids in various proteins and biological products
  • Mild reaction conditions
  • Short reaction time
  • High nucleic acid removal efficiency