Learn More
Sigma Aldrich Fine Chemicals Biosciences Nuclei Isolation Kit: Nuclei PURE Prep sufficient for 15 nuclei preparations (~1-10x107 cells or 1g of tissue per preparation) | 1KT
Catalog No. NC2213511
Encompass
Nuclei Isolation Kit: Nuclei PURE Prep sufficient for 15 nuclei preparations (~1-10x107 cells or 1g of tissue per preparation) | 1KT
This item is not returnable. View return policy
Catalog No. NC2213511 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NUC2011KT
Nuclei Isolation Kit: Nuclei PURE Prep sufficient for 15 nuclei preparations (~1-10x107 cells or 1g of tissue per preparation) | 1KT