Learn More
Sigma Aldrich Fine Chemicals Biosciences Nuclei Isolation Kit: Nuclei EZ Prep
Catalog No. NC1826863
Encompass
The Nuclei EZ Prep Kit was developed for the rapid isolation of nuclei from mammalian cells. The kit provides a high yield of nuclei from commonly used cell types. Suitable as a source of nuclear components, to produce nuclei for in vitro apoptosis assays, and for functional studies.
Catalog No. NC1826863 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. NUC1011KT
The Nuclei EZ Prep Kit was developed for the rapid isolation of nuclei from mammalian cells. The kit provides a high yield of nuclei from commonly used cell types. Suitable as a source of nuclear components, to produce nuclei for in vitro apoptosis assays, and for functional studies.