Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Nucleic Acid Detection Blocking Buffer

Catalog No. FER89880A Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FER89880A 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FER89880A Supplier Thermo Scientific™ Supplier No. 89880A
Get 16% Off Protein Research Essentials
Use promo code PROTEIN16
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific™ Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component.

Nucleic Acid Detection Blocking Buffer is intended for use with the Thermo Scientific™ Chemiluminescent Nucleic Acid Detection Module Kit, of which it is also a component. Non-specific binding of probe to membrane can be minimized by incubating the membrane in Blocking Buffer for 15 minutes.

Related Products

  • Chemiluminescent Nucleic Acid Detection Module Kit (Cat. No. 89880)

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at 4°C.
Buffer Blocking Buffer
Product Type Nucleic Acid Detection Blocking Buffer
Quantity 500 mL

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.