Learn More
Cayman Chemical NucleIc AcId Dye Green I 500uL
SDP
Catalog No. 502911229
Encompass_Preferred
A multivalent ionizable cationic lipid has been used in the generation of liposomes complexed with plasmid DNA for use in vitro and in vivo has been used in combination with DC-Chol and 12-DOPE in the generation of lipid nanocarriers for siRNA delivery to A549 cells GL67-containing liposomes in complex with plasmid DNA for the gene encoding CFTR leads to the expression of functional wild-type CFTR in HEK293 cells aerosol administration of GL67-containing liposomes in complex with plasmid DNA for the gene encoding CFTR increases trachea and whole lung mRNA levels of CFTR
Catalog No. 50-291-1229 Supplier Cayman Chemical Supplier No. 35669500 UL
A multivalent ionizable cationic lipid has been used in the generation of liposomes complexed with plasmid DNA for use in vitro and in vivo has been used in combination with DC-Chol and 12-DOPE in the generation of lipid nanocarriers for siRNA delivery to A549 cells GL67-containing liposomes in complex with plasmid DNA for the gene encoding CFTR leads to the expression of functional wild-type CFTR in HEK293 cells aerosol administration of GL67-containing liposomes in complex with plasmid DNA for the gene encoding CFTR increases trachea and whole lung mRNA levels of CFTR