Learn More
Epigentek Inc Nucleic Acid Isolation Enhancer (Glycogen Solution) (500 ul)
SDP
Catalog No. 501096839
Encompass_Preferred
The Nucleic Acid Isolation Enhancer is used as a carrier for nucleic acid precipitation in ethanol.
Catalog No. 50-109-6839 Supplier Epigentek Inc Supplier No. R10011
The Nucleic Acid Isolation Enhancer is used as a carrier for nucleic acid precipitation in ethanol.