Learn More
AB Sciex LLC Nucleic Acid Extended Range Gel Multi Pack
Catalog No. NC2892662
Encompass
A 4-pack of the Nucleic Acid Extended Range Gel, which is found in the RNA 9000 Purity & Integrity kit. This gel pack allows for the purchasing of only the gel and the ability to use a dye and ladder of your choosing.
Catalog No. NC2892662 Supplier AB Sciex LLC Supplier No. 5308349
A 4-pack of the Nucleic Acid Extended Range Gel, which is found in the RNA 9000 Purity & Integrity kit. This gel pack allows for the purchasing of only the gel and the ability to use a dye and ladder of your choosing.