Learn More
Fluidigm Corp NUCLEIC ACID INTERCALATOR, NAT
SDP
Catalog No. NC0533143
Encompass
Cell-ID Intercalator-Ir is a cationic nucleic acid intercalator that contains natural-abundance iridium (191Ir and 193Ir), which is used in CyTOF analysis for discrimination of dead cells from live cells (if staining is done before cell fixation) or to discriminate single nucleated cells from doublets (if staining is done post-fixation). It is a live cell membrane-impermeable dye.
Catalog No. NC0533143 Supplier Fluidigm Corp Supplier No. 201192B
Cell-ID Intercalator-Ir is a cationic nucleic acid intercalator that contains natural-abundance iridium (191Ir and 193Ir), which is used in CyTOF analysis for discrimination of dead cells from live cells (if staining is done before cell fixation) or to discriminate single nucleated cells from doublets (if staining is done post-fixation). It is a live cell membrane-impermeable dye.