Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Edvotek™ Nucleic Acid Testing for COVID-19
SDPSCI_ED

Catalog No. S52001 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
S52001 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S52001 Supplier Edvotek™ Supplier No. 123
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

In this simulated medical test, electrophoresis is used to detect the presence of the SARS-CoV-2 virus in samples from patients with symptoms of COVID-19.

  • Hands-on simulation of molecular testing for COVID-19
  • Students learn about the characteristics of coronaviruses
  • Electrophoresis is used to analyze simulated RT-PCR samples
  • Complete experiment in 45 minutes
  • For 8 Gels
  • Kit includes Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), Practice Gel Loading Solution, FlashBlue™ DNA Stain, & Disposable Pipets
  • Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended

Specifications

Grade 8 to Undergraduate
Age 14 to 20
For Use With (Application) Simulated COVID-19 lab experiment
Includes Instructions, Ready-to-Load QuickStripTM DNA Samples, UltraSpec-AgaroseTM, Electrophoresis Buffer (50 X), Practice Gel Loading Solution, FlashBlueTM DNA Stain, & Disposable Pipets
Content And Storage Room Temperature Stable. Storage of Ready-to-Load QuickStripTM samples in the Refrigerator is Recommended
Class Size 8 Lab Groups
Product Type Electrophoreseis Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.