Learn More
Alpco Ltd Nucleo-h IgG ELISA
SDP
Supplier:  Alpco Ltd 35NUCHUE01
Encompass_Preferred
The Nucleo-h IgG ELISA is a solid phase enzyme immunoassay with human nativenucleosomes isolated from the eukaryotic cell line HeLa for the quantitative and qualitativedetection of antibodies against nucleosomes and their components dsDNA and histones in human serum. The Nucleo-h IgG ELISA is for Research Use Only. The Nucleo-h IgG ELISA is not for use in diagnostic procedures.
Catalog No. NC2329142