Learn More
Macherey-Nagel NucleoBond Finalizer Large Plus, 20 finalizers
Catalog No. NC9941269
Encompass_Preferred
NucleoBond Finalizer Large Plus (20 sets) filters for plasmid DNA precipitation, to be used with NucleoBond Xtra Maxi kits - 20 NucleoBond Finalizers Large, 20 syringes of 30 mL, and 20 syringes of 1 mL
Catalog No. NC9941269 Supplier Macherey-Nagel Supplier No. 740419.20
NucleoBond Finalizer Large Plus (20 sets) filters for plasmid DNA precipitation, to be used with NucleoBond Xtra Maxi kits - 20 NucleoBond Finalizers Large, 20 syringes of 30 mL, and 20 syringes of 1 mL