Learn More
Macherey-Nagel NucleoBond Finalizer Plus, 20 finalizers
Catalog No. NC1620155
Encompass_Preferred
NucleoBond Finalizer Plus (20) filters for plasmid DNA precipitation, to be used with NucleoBond PC 100, PC 500, and Xtra Midi(EF) kits - 20 NucleoBond Finalizers, 20 syringes of 30 mL, and 20 syringes of 1 mL
Catalog No. NC1620155 Supplier Macherey-Nagel Supplier No. 740520.20
NucleoBond Finalizer Plus (20) filters for plasmid DNA precipitation, to be used with NucleoBond PC 100, PC 500, and Xtra Midi(EF) kits - 20 NucleoBond Finalizers, 20 syringes of 30 mL, and 20 syringes of 1 mL