Learn More
Macherey-Nagel MACHEREY-NAGEL NucleoBond HMW DNA Kit, High Integrity DNA Purification, Enzymatic Lysis Included, 20 Preps
Supplier:  Macherey-Nagel 740160.20
Encompass_Preferred
MACHEREY-NAGEL NucleoBond HMW DNA kit is designed for the purification of high molecular weight DNA, ensuring high integrity suitable for long-read and Next Generation Sequencing applications. The kit utilizes anion-exchange chromatography and includes Proteinase K for gentle enzymatic lysis, minimizing DNA shearing. It is ideal for isolating DNA from cultured cells, tissue, and plant samples, making it a versatile choice for demanding research environments.
Catalog No. NC2099853