Learn More
Macherey-Nagel NucleoBond PC 10000 EF (5)
Catalog No. NC0389435
Encompass_Preferred
NucleoBond PC 10000 EF (5) 5 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond AX 10000 Columns, NucleoBond Bottle Top Filters Type 2, buffers, RNase A
Catalog No. NC0389435 Supplier Macherey-Nagel Supplier No. 740548
NucleoBond PC 10000 EF (5) 5 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond AX 10000 Columns, NucleoBond Bottle Top Filters Type 2, buffers, RNase A