Learn More
Macherey-Nagel NucleoBond Smart Rack
Catalog No. NC1477672
Encompass_Preferred
NucleoBond Smart Rack light grey rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 20, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1
Catalog No. NC1477672 Supplier Macherey-Nagel Supplier No. 740413
NucleoBond Smart Rack light grey rack for use with NucleoBond Xtra Midi, Xtra Maxi, Xtra BAC, AX 20, AX 100, AX 500, AX 2000, and AX 10000 Columns pack of 1