Learn More
Macherey-Nagel NucleoBond Xtra EF Buffer Set I
Catalog No. NC1488639
Encompass_Preferred
NucleoBond Xtra EF Buffer Set I buffer set for the isolation of endotoxin-free low-copy plasmids, BACs, PACs, and P1 constructs, only applicable with NucleoBond Xtra EF kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES-EF, Buffer LYS-EF, Buffer NEU-EF, RNase A
Catalog No. NC1488639 Supplier Macherey-Nagel Supplier No. 740427
NucleoBond Xtra EF Buffer Set I buffer set for the isolation of endotoxin-free low-copy plasmids, BACs, PACs, and P1 constructs, only applicable with NucleoBond Xtra EF kits, sufficient for 12 Xtra Maxi preps or 18 Xtra Midi preps - Buffer RES-EF, Buffer LYS-EF, Buffer NEU-EF, RNase A