Learn More
Macherey-Nagel MACHEREY-NAGEL NucleoBond Xtra Midi Columns, High Yield Ultrapure DNA, Fast 30 Min Prep, Package of 10 Columns
Catalog No. NC2337633
Encompass_Preferred
MACHEREY-NAGEL NucleoBond Xtra Midi Columns offer a rapid solution for isolating high-quality plasmid DNA through advanced anion-exchange chromatography. With integrated filters and a streamlined workflow, users can achieve ultrapure yields in approximately 30 minutes, significantly outperforming competitors. The unique silica-based resin ensures specific binding of plasmid DNA, making it ideal for sensitive applications like transfection and sequencing.
Catalog No. NC2337633 Supplier Macherey-Nagel Supplier No. 740410.10S
MACHEREY-NAGEL NucleoBond Xtra Midi Columns offer a rapid solution for isolating high-quality plasmid DNA through advanced anion-exchange chromatography. With integrated filters and a streamlined workflow, users can achieve ultrapure yields in approximately 30 minutes, significantly outperforming competitors. The unique silica-based resin ensures specific binding of plasmid DNA, making it ideal for sensitive applications like transfection and sequencing.