Learn More
Macherey-Nagel NucleoBond Xtra Midi Plus EF, 50 preps
Catalog No. NC1270087
Encompass_Preferred
NucleoBond Xtra Midi Plus EF (50) 50 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond Xtra Midi Columns, NucleoBond Xtra Midi Column Filters, NucleoBond Finalizers, syringes, buffers, RNase A
Catalog No. NC1270087 Supplier Macherey-Nagel Supplier No. 740422.50
NucleoBond Xtra Midi Plus EF (50) 50 preps for the isolation of endotoxin-free plasmid DNA - NucleoBond Xtra Midi Columns, NucleoBond Xtra Midi Column Filters, NucleoBond Finalizers, syringes, buffers, RNase A