Learn More
Lonza Walkersville Cell Line Nucleofector Kit C
Catalog No. NC9535446
Encompass
The Cell Line Nucleofector Kit C is one of our five Nucleofector Kits suited for transfecting cell lines in the Nucleofector 2b Device. This kit has been determined to be optimal for transfecting following cell types:A2058CCRF-CEMMDA-MB-453MEG-01MG-63NCI-H1299 [H1299]NS0 (ECACC)SK-BR-3T2U266B1U-937The respective cell type-specific Optimized Protocol or recommendation is available under "Instructions" or in our knowledge database. In this Optimized Protocol the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of this cell type.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution C0.5 mL Supplement 230 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes
Catalog No. NC9535446 Supplier Lonza Walkersville Supplier No. VCA1004
The Cell Line Nucleofector Kit C is one of our five Nucleofector Kits suited for transfecting cell lines in the Nucleofector 2b Device. This kit has been determined to be optimal for transfecting following cell types:A2058CCRF-CEMMDA-MB-453MEG-01MG-63NCI-H1299 [H1299]NS0 (ECACC)SK-BR-3T2U266B1U-937The respective cell type-specific Optimized Protocol or recommendation is available under "Instructions" or in our knowledge database. In this Optimized Protocol the best Nucleofection conditions are indicated. In addition, we share our experience and knowledge for treatment of this cell type.Benefits:High transfection efficiencies in difficult-to-transfect cell linesApplications:Transfection of cell numbers in the range of 2 x 10⁵ to 2 x 10⁷ cells Low-throughput transfection in single cuvette formatContent:2.25 mL Nucleofector Solution C0.5 mL Supplement 230 µg pmaxGFP Vector (0.5 μg/μl in 10 mM Tris pH 8.0)25 Aluminum cuvette (100 µL)25 Single use pipettes