Learn More
Lonza Walkersville Basic Nucleofector Kit for Primary Mammalian Smooth Muscle Cells
Catalog No. NC9667210
Encompass
In addition to various cell type-specific kits for primary smooth muscle cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian smooth muscle cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for smooth muscle cells. Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support TeamTransfection efficiencies up to 95%Viabilities up to 96%Applications:Kit suited for smooth muscle cells from different mammalian species and various organs Already tested for porcine vascular smooth muscle cells and coronary artery smmoth muscle cells (CASMC)
Catalog No. NC9667210 Supplier Lonza Walkersville Supplier No. VPI1004
In addition to various cell type-specific kits for primary smooth muscle cells, we also provide a cell group-specific Basic Nucleofector Kit. This kit is suited to determine the optimal transfection conditions for any mammalian smooth muscle cell type that lacks a cell type specific Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). In a first experiment a selection of different Nucleofector Programs is tested in combination with the Basic Nucleofector Solution for smooth muscle cells. Benefits:Optimization can be performed within 1 experiment Detailed protocols guiding through the optimization procedure Optional result fine tuning with help of our Scientific Support TeamTransfection efficiencies up to 95%Viabilities up to 96%Applications:Kit suited for smooth muscle cells from different mammalian species and various organs Already tested for porcine vascular smooth muscle cells and coronary artery smmoth muscle cells (CASMC)