Learn More
Lonza Walkersville Cell Line Optimization Nucleofector Kit for Nucleofector Device
Catalog No. NC9446456
Encompass
The Cell Line Optimization Nucleofector Kit is the ideal tool to determine the optimal transfection conditions for any difficult-to-transfect cell line that lacks an Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). It enables you to conveniently determine the optimal Nucleofection condition of your cell line of interest within one experiment. The kit contains two different Cell Line Nucleofector Solutions, V and L, each of which should be tested in combination with seven different Nucleofector Programs. It is suitable for 18 optimization reactions. Fine-tuning for optimal results can then be performed together with our Scientific Support Team.Benefits:Efficient transfection of virtually any difficult-to-transfect cell lineSimple and rapid optimization completed within just one experimentApplications:Optimization of Nucleofection conditions of virtually any difficult-to-transfect cell line with the Nucleofector II/2b Device
Catalog No. NC9446456 Supplier Lonza Walkersville Supplier No. VCO1001N
The Cell Line Optimization Nucleofector Kit is the ideal tool to determine the optimal transfection conditions for any difficult-to-transfect cell line that lacks an Optimized Protocol for the Nucleofector 2b Device (or older generations I or II). It enables you to conveniently determine the optimal Nucleofection condition of your cell line of interest within one experiment. The kit contains two different Cell Line Nucleofector Solutions, V and L, each of which should be tested in combination with seven different Nucleofector Programs. It is suitable for 18 optimization reactions. Fine-tuning for optimal results can then be performed together with our Scientific Support Team.Benefits:Efficient transfection of virtually any difficult-to-transfect cell lineSimple and rapid optimization completed within just one experimentApplications:Optimization of Nucleofection conditions of virtually any difficult-to-transfect cell line with the Nucleofector II/2b Device