Learn More
Enzo Life Sciences NUCLEOLAR-ID Green detection kit (1Kit)
SDP
Supplier:  Enzo Life Sciences ENZ51009500
Encompass_Preferred
Enzo  Life Sciences’ NUCLEOLAR-ID Green Detection Kit contains a proprietary dye suitable for live-cell staining of nucleoli. The dye allows examination of nucleolar dynamic changes in intracellular distribution, trafficking and localization arising from biological processes such as the cell cycle and ribosome biogenesis. The kit is compatible with most fluorescence detection systems, including conventional and confocal fluorescence microscopes, as well as High Content Screening (HCS) platforms. This kit is specifically designed for visualizing nucleoli in living cells. The dye in the kit is resistant to photobleaching, facilitating its use in imaging applications. This nucleolar dye may be used in conjunction with common live-cell nuclear counterstains such as Hoechst 33342, Draq5™, Vybrant DyeCycle™ Ruby and Enzo’s NUCLEAR-ID Red dye.Applications: Fluorescence microscopy, Fluorescent detection .
Catalog No. 50-201-2584