Learn More
Aat Bioquest Nucleolus Red™ DCS1 *Dead Cell Staining*, 100 Tests
SDP
Catalog No. NC3728108
Encompass_Preferred
Nucleolus Red™ DCS1 is a cell-impermeant fluorescent dye that selectively binds to RNA within the nucleolus, a non-membrane-bound nuclear compartment responsible for ribosomal RNA (rRNA) transcription and processing. Upon binding to RNA, the dye exhibits strong red fluorescence, whereas its interaction with DNA results in only weak fluorescence, providing exceptional specificity for nucleolar RNA detection.
Catalog No. NC3728108 Supplier Aat Bioquest Supplier No. 17684
Nucleolus Red™ DCS1 is a cell-impermeant fluorescent dye that selectively binds to RNA within the nucleolus, a non-membrane-bound nuclear compartment responsible for ribosomal RNA (rRNA) transcription and processing. Upon binding to RNA, the dye exhibits strong red fluorescence, whereas its interaction with DNA results in only weak fluorescence, providing exceptional specificity for nucleolar RNA detection.