Learn More
Macherey-Nagel NucleoMag RNA, 1 x 96 preps
Catalog No. NC1729154
Encompass_Preferred
NucleoMag RNA (1 x 96) 1 x 96 preps for the purification of total RNA - NucleoMag B-Beads, Buffers, Reducing Agent TCEP, RNase-free rDNase
Catalog No. NC1729154 Supplier Macherey-Nagel Supplier No. 744350.1
NucleoMag RNA (1 x 96) 1 x 96 preps for the purification of total RNA - NucleoMag B-Beads, Buffers, Reducing Agent TCEP, RNase-free rDNase