Learn More
ABclonal Technology Nucleophosmin Rabbit mAb
SDP
Catalog No. 5000010329
Encompass_Preferred
The protein encoded by this gene is involved in several cellular processes including centrosome duplication protein chaperoning and cell proliferation The encoded phosphoprotein shuttles between the nucleolus nucleus and cytoplasm chaperoning ribosomal proteins and core histones from the nucleus to the cytoplasm This protein is also known to sequester the tumor suppressor ARF in the nucleolus protecting it from degradation until it is needed Mutations in this gene are associated with acute myeloid leukemia Dozens of pseudogenes of this gene have been identified
Catalog No. 50-000-10329 Supplier ABclonal Technology Supplier No. A2729720UL
The protein encoded by this gene is involved in several cellular processes including centrosome duplication protein chaperoning and cell proliferation The encoded phosphoprotein shuttles between the nucleolus nucleus and cytoplasm chaperoning ribosomal proteins and core histones from the nucleus to the cytoplasm This protein is also known to sequester the tumor suppressor ARF in the nucleolus protecting it from degradation until it is needed Mutations in this gene are associated with acute myeloid leukemia Dozens of pseudogenes of this gene have been identified