Learn More
Bioss Nucleoporin p62 - N-terminal r
SDP
Catalog No. 5000066387
Encompass_Preferred
Nucleoporin p62 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB M436Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol
Catalog No. 50-000-66387 Supplier Bioss Supplier No. BSM70579M
Nucleoporin p62 (N-terminal region) Primary Antibody Unconjugated Host Species Mouse Target Species Human Mouse Rat Chicken Clonality Monoclonal Applications WB M436Concentration Size 100 ul Storage Buffer PBS 1 mg/ml BSA 0 05% NaN3 and 50% glycerol