Learn More
Medchemexpress LLC Nucleoprotein/NP protein, H1N1 (I116M, AAM75159, sf9, His) | >95.0% | 50 UG
SDP
Catalog No. 5000294325
Encompass_Preferred
Recombinant Influenza A (H1N1) nucleoprotein containing the I116M substitution, expressed in Sf9 insect cells and purified with a C-terminal His tag. Supplied lyophilized and intended for research applications such as immunoassays, antibody development, and structural studies.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • C-terminal His tag for straightforward purification and detection.
  • Contains I116M substitution for sequence-specific studies.
  • >95.0% purity determined by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for long-term storage.
  • Stable when stored at -20°C; freeze aliquots for extended stability.
  • Suitable for ELISA, antibody generation, and structural analysis.
Catalog No. 50-002-94325 Supplier Medchemexpress LLC Supplier No. HYP7467650UG
Recombinant Influenza A (H1N1) nucleoprotein containing the I116M substitution, expressed in Sf9 insect cells and purified with a C-terminal His tag. Supplied lyophilized and intended for research applications such as immunoassays, antibody development, and structural studies.
  • Expressed in Sf9 insect cells for eukaryotic folding.
  • C-terminal His tag for straightforward purification and detection.
  • Contains I116M substitution for sequence-specific studies.
  • >95.0% purity determined by reducing SDS-PAGE.
  • Lyophilized formulation with stabilizers for long-term storage.
  • Stable when stored at -20°C; freeze aliquots for extended stability.
  • Suitable for ELISA, antibody generation, and structural analysis.