Learn More
Diagnocine LLC NUCLEOSEEING-LIVE NUCLEUSGREEN
SDP
Catalog No. NC1727038
Encompass
NucleoSeeing .Storage: -20°CSize: 0.1mgNucleoSeeing is a novel live cell imaging probe which emits green fluorescence by binding to DNA specifically.NucleoSeeing can be used for not only in animal cells or tissues, but also Guard cell of Arabidopsis thaliana with high S/N ratio. Moreover, NucleoSeeing can be used as a pH sensor in nucleus.
Catalog No. NC1727038 Supplier Diagnocine LLC Supplier No. FNKFDV0029
NucleoSeeing .Storage: -20°CSize: 0.1mgNucleoSeeing is a novel live cell imaging probe which emits green fluorescence by binding to DNA specifically.NucleoSeeing can be used for not only in animal cells or tissues, but also Guard cell of Arabidopsis thaliana with high S/N ratio. Moreover, NucleoSeeing can be used as a pH sensor in nucleus.