Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Nucleoside-analog-1 | 876707-99-2 | 95.0% | 267.20 | C9H9N5O5 | 10 MG
SDP

Catalog No. 5000299273
Encompass_Preferred

An azide-functionalized nucleoside reagent for click chemistry and bioconjugation that reacts with alkynes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and with strained alkynes via strain-promoted azide-alkyne cycloaddition (SPAAC). Intended for research use.

  • Azide-functionalized nucleoside for bioconjugation.
  • Reacts in CuAAC and SPAAC click chemistry.
  • Molecular weight 267.20, formula C9H9N5O5.
  • Purity ≥95.0%.
  • White to off-white solid; powder storage stable at -20°C.
  • Available in small research pack sizes, including 10 MG.

Catalog No. 50-002-99273 Supplier Medchemexpress LLC Supplier No. HY7765110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

An azide-functionalized nucleoside reagent for click chemistry and bioconjugation that reacts with alkynes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and with strained alkynes via strain-promoted azide-alkyne cycloaddition (SPAAC). Intended for research use.

  • Azide-functionalized nucleoside for bioconjugation.
  • Reacts in CuAAC and SPAAC click chemistry.
  • Molecular weight 267.20, formula C9H9N5O5.
  • Purity ≥95.0%.
  • White to off-white solid; powder storage stable at -20°C.
  • Available in small research pack sizes, including 10 MG.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.